Order Entry
Belgium
ContactUsLinkComponent
D-(-)-Mannitol 97.0-102.0% (dry basis), powder USP, Multi-Compendial GMP Manufactured Product, J.T.Baker®
D-(-)-Mannitol 97.0-102.0% (dry basis), powder USP, Multi-Compendial GMP Manufactured Product, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
D-(-)-Mannitol 97.0-102.0% (dry basis), powder USP, Multi-Compendial GMP Manufactured Product, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    97.0 - 102.0%
  • Meets E.P. Chemical Specifications:
    Passes Test
  • Meets U.S.P Requirements:
    ≤2.0%
  • Meets B.P. Chemical Specifications:
    ≤2.0%
  • Meets J.P. Chemical Specifications:
    ≤0.10%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤2.0%
  • Bulk Pharmaceutical Chemical:
    ≤0.5%
  • USP - Assay (dried basis):
    97.0 - 102.0 %
  • USP - Identification:
    Passes Test
  • USP - Related Substances: Sorbitol:
    ≤ 2.0 %
  • USP - Related Substances: Sum of isomalt& maltitol:
    ≤ 2.0 %
  • USP - Related Substances: Unspecified impurities:
    ≤ 0.10 %
  • USP - Related Substances: Total impurities:
    ≤ 2.0 %
  • USP - Loss on Drying at 105°C:
    ≤ 0.5 %
  • USP - Melting Point:
    165 - 170 °C
  • USP - Reducing Sugars:
    ≤ 0.1 %
  • USP - Nickel (Ni):
    ≤ 1 ppm
  • USP - Appearance of Solution:
    Passes Test
  • USP - Conductivity (uS cm-1):
    ≤ 20 µS/cm
  • USP - Total Aerobic Microbial Count:
    ≤ 100 cfu/g
  • USP - Total Yeast and Mold Count:
    ≤ 100 cfu/g
  • USP - Escherichia coli:
    Passes Test
  • USP - Bacterial Endotoxins, IU/g:
    ≤ 2.5
  • EP/BP - Assay (as HOCH₂(CHOH)₄CH₂OH) (dried basis):
    97.0 - 102.0 %
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Melting Point:
    165 - 170 °C
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Conductivity, uS cm-1:
    ≤ 20
  • EP/BP - Reducing Sugars:
    ≤ 0.1 %
  • EP/BP - Related Substances: Impurity A:
    ≤ 2.0 %
  • EP/BP - Related Substances: Sum of Impurities B & C:
    ≤ 2.0 %
  • EP/BP - Related Substances: Unspecified Impurities, each:
    ≤ 0.10 %
  • EP/BP - Related Substances: Total Impurities:
    ≤ 2.0 %
  • EP/BP - Loss on Drying:
    ≤ 0.5 %
  • EP/BP - Escherichia coli:
    None Detected
  • EP/BP - Salmonella:
    None Detected
  • EP/BP - Total Yeast and Mold Count:
    ≤ 100 cfu/g
  • EP/BP - Total Aerobic Microbial Count:
    ≤ 100 cfu/g
  • EP/BP Endotoxin Concentration, <2.5 IU/g:
    Passes Test
  • JP - Assay (dried basis):
    97.0 - 102.0 %
  • JP - Identification:
    Passes Test
  • JP - Related Substances: Sorbitol:
    ≤ 2.0 %
  • JP - Related Substances: Sum of isomalt & maltitol:
    ≤ 2.0 %
  • JP - Related Substances: Unspecified impurities:
    ≤ 0.10 %
  • JP - Related Substances: Total impurities:
    ≤ 2.0 %
  • JP - Heavy Metals (as Pb):
    ≤ 5 ppm
  • JP - Loss on Drying at 105°C:
    ≤ 0.5 %
  • JP - Melting Point:
    165 - 170 °C
  • JP - Glucose:
    ≤ 0.1 %
  • JP - Nickel (Ni):
    ≤ 1 ppm
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Conductivity (uS cm-1):
    ≤ 20 µS/cm

Specification Test Results

Related Information

Frequently Bought Together