Orders LinkContactUsLinkComponent
Autoclavable goggles
Autoclavable goggles
Supplier:   UNIVET
Autoclavable goggles
Supplier:  UNIVET
Certificates
Autoclavable goggles
4 Options Available Below
Product Details & Documents
The autoclavable goggles are designed for use in rooms with controlled contaminations or in aseptic conditions.
  • Available with direct or indirect ventilation
  • Elastic band in anti-static silicone with quick and smooth system
  • RX insert
  • Goggles with polycarbonate lens reusable up to 40 autoclave
The soft structure supports multiple cycles of steam, gamma or beta sterilisation without perceptible changes. The autoclavable goggles has a polycarbonate lens with anti-fog coating AF121 and anti-scratch.
Certifications: EN 166.
Delivery information: Available with correction insert and replacement lens.
Product Family Options
Product Information
  • Type
    Lenses
    Colour
    Availability
    Price
  • 111-2349
    Cleanroom goggles
    Cleanroom goggles
    Directly vented
    Clear βγoxene® disc
    Blue
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 293,00 €
    293,00 €
    1 * 5 ST
       293,00 €
  • 111-2428
    Cleanroom goggles
    Cleanroom goggles
    Anti-UV, anti-fog, autoclavable, directly vented
    Clear PC
    Blue
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 357,00 €
    357,00 €
    1 * 5 ST
       357,00 €
  • 111-2429
    Cleanroom goggles
    Cleanroom goggles
    Anti-UV, autoclavable, indirectly vented
    Clear PC
    Blue
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 293,00 €
    293,00 €
    1 * 5 ST
       293,00 €
  • 111-2430
    Cleanroom goggles
    Cleanroom goggles
    Anti-UV, anti-fog, autoclavable, indirectly vented
    Clear PC
    Blue
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 366,00 €
    366,00 €
    1 * 5 ST
       366,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare lens for 611S, clear PC
    Spare lens for 611S, clear PC
    UNVT042.060
    Log in for availability
    1 * 1 ST - 14,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Head band, silicone, 700×30×1,4 mm
    Head band, silicone, 700×30×1,4 mm
    UNVT3PA299
    Log in for availability
    1 * 1 ST - 19,60 €
Product Information
  • Catalog #
    Availability
    Price
  • RX optical insert for goggles
    RX optical insert for goggles
    UNVT601.OP.00.99
    Log in for availability
    1 * 1 ST - 14,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare lens, clear PC
    Spare lens, clear PC
    UNVT042.120M
    Log in for availability
    1 * 1 ST - 33,40 €