Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ganciclovir, Reference Standards

Ganciclovir, Reference Standards

Supplier:   USP
Ganciclovir, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1288306.
  • 1288317.
  • 1288339.
  • Description:
  • Ganciclovir
  • Ganciclovir Related Compound A
  • Ganciclovir Mono-N-methyl Valinate
  • Synonym(s):
  • (RS)-2-Amino-9-(2,3-dihydroxy-propoxymethyl)-1,9-dihydro-purin-6-one
  • 2-(RS)-[(guanin-9-yl)methoxy]-3-hydroxypropyl N-methyl-L-valinate hydrochloride
  • CAS No.:
  • 82410-32-0
  • 86357-09-7
  • 1401562-16-0
  • Formula:
  • C₉H₁₃N₅O₄
  • C₉H₁₃N₅O₄
  • C₁₅H₂₅ClN₆O₅

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1288306.
    Ganciclovir
    Ganciclovir
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 823,00 €
    823,00 €
    1 * 200 mg
       823,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1288306.
    Description
    Ganciclovir
    Synonym(s)
    CAS No.
    82410-32-0
    Formula
    C₉H₁₃N₅O₄
  • 1288317.
    Ganciclovir Related Compound A
    Ganciclovir Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 660,00 €
    1 660,00 €
    1 * 15 mg
       1 660,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1288317.
    Description
    Ganciclovir Related Compound A
    Synonym(s)
    (RS)-2-Amino-9-(2,3-dihydroxy-propoxymethyl)-1,9-dihydro-purin-6-one
    CAS No.
    86357-09-7
    Formula
    C₉H₁₃N₅O₄
  • 1288339.
    Ganciclovir Mono-N-methyl Valinate
    Ganciclovir Mono-N-methyl Valinate
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 250,00 €
    1 250,00 €
    1 * 10 mg
       1 250,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1288339.
    Description
    Ganciclovir Mono-N-methyl Valinate
    Synonym(s)
    2-(RS)-[(guanin-9-yl)methoxy]-3-hydroxypropyl N-methyl-L-valinate hydrochloride
    CAS No.
    1401562-16-0
    Formula
    C₁₅H₂₅ClN₆O₅