Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flutamide, Reference Standards

Flutamide, Reference Standards

Supplier:   USP
Flutamide, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1285851.
  • 1285862.
  • 1285986.
  • 1285997.
  • Description:
  • Flutamide
  • o-Flutamide
  • Flutamide Related Compound B
  • Flutamide Related Compound A
  • Synonym(s):
  • N-[4-nitro-3-(trifluoromethyl)phenyl]acetamide
  • 4-nitro-3-(trifluoromethyl)aniline
  • CAS No.:
  • 13311-84-7
  • 151262-93-0
  • 393-12-4
  • 393-11-3
  • Formula:
  • C₁₁H₁₁F₃N₂O₃
  • C₁₁H₁₁F₃N₂O₃
  • UN No.:
  • 3261

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1285851.
    Flutamide
    Flutamide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 375,00 €
    375,00 €
    1 * 200 mg
       375,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1285851.
    Description
    Flutamide
    Synonym(s)
    CAS No.
    13311-84-7
    Formula
    C₁₁H₁₁F₃N₂O₃
    UN No.
  • 1285862.
    o-Flutamide
    o-Flutamide
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 050,00 €
    1 050,00 €
    1 * 50 mg
       1 050,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1285862.
    Description
    o-Flutamide
    Synonym(s)
    CAS No.
    151262-93-0
    Formula
    C₁₁H₁₁F₃N₂O₃
    UN No.
  • 1285986.
    Flutamide Related Compound B
    Flutamide Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 260,00 €
    1 260,00 €
    1 * 20 mg
       1 260,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1285986.
    Description
    Flutamide Related Compound B
    Synonym(s)
    N-[4-nitro-3-(trifluoromethyl)phenyl]acetamide
    CAS No.
    393-12-4
    Formula
    UN No.
    3261
  • 1285997.
    Flutamide Related Compound A
    Flutamide Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 300,00 €
    1 300,00 €
    1 * 20 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1285997.
    Description
    Flutamide Related Compound A
    Synonym(s)
    4-nitro-3-(trifluoromethyl)aniline
    CAS No.
    393-11-3
    Formula
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.