Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Meloxicam, Reference Standards

 :   USP
Meloxicam, Reference Standards
 :  USP
 
Meloxicam, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1379401.
  • 1379412.
  • 1379423.
  • 1379434.
  • Description:
  • Meloxicam
  • Meloxicam Related Compound A
  • Meloxicam Related Compound B
  • Meloxicam Related Compound C
  • Synonym(s):
  • ethyl-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate- 1,1-dioxide
  • 5-Methylthiazol-2-amine
  • Isopropyl 4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide
  • CAS No.:
  • 71125-38-7
  • 24683-26-9
  • 7305-71-7
  • 118854-48-1
  • Formula:
  • C₁₄H₁₃N₃O₄S₂
  • C₁₂H₁₃NO₅S
  • C₄H₆N₂S
  • C₁₃H₁₅NO₅S
  • UN No.:
  • 2811
  • 3077

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Meloxicam
    USP reference standard
    Vial
    400 mg
       
       
    1 * 400 mg
    1 * 400 mg - 646,00 €
    646,00 €
    1 * 400 mg
       646,00 €

     
     
    Specifications
    Cat. No.
    1379401.
    Description
    Meloxicam
    Synonym(s)
    CAS No.
    71125-38-7
    Formula
    C₁₄H₁₃N₃O₄S₂
    UN No.
    2811
  • Meloxicam Related Compound A
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 300,00 €
    1 300,00 €
    1 * 25 mg
       1 300,00 €

     
     
    Specifications
    Cat. No.
    1379412.
    Description
    Meloxicam Related Compound A
    Synonym(s)
    ethyl-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate- 1,1-dioxide
    CAS No.
    24683-26-9
    Formula
    C₁₂H₁₃NO₅S
    UN No.
  • Meloxicam Related Compound B
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 260,00 €
    1 260,00 €
    1 * 25 mg
       1 260,00 €

     
     
    Specifications
    Cat. No.
    1379423.
    Description
    Meloxicam Related Compound B
    Synonym(s)
    5-Methylthiazol-2-amine
    CAS No.
    7305-71-7
    Formula
    C₄H₆N₂S
    UN No.
    3077
  • Meloxicam Related Compound C
    USP reference standard
    Vial
    30 mg
       
       
    1 * 30 mg
    1 * 30 mg - 1 360,00 €
    1 360,00 €
    1 * 30 mg
       1 360,00 €

     
     
    Specifications
    Cat. No.
    1379434.
    Description
    Meloxicam Related Compound C
    Synonym(s)
    Isopropyl 4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide
    CAS No.
    118854-48-1
    Formula
    C₁₃H₁₅NO₅S
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.