Orders LinkContactUsLinkComponent
pH indicator strips, Whatman™
pH indicator strips, Whatman™
pH indicator strips, Whatman™
Certificates
pH indicator strips, Whatman™
pH indicator strips, Whatman™
2 Options Available Below
Product Details & Documents
Universal indicator strips which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
Product Family Options
Product Information
  • pH range
    Availability
    Price
  • 700-0442
    Universal indicator papers, non bleeding strips
    Universal indicator papers, non bleeding strips
    0 - 14,0
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 22,60 €
    22,60 €
    1 * 100 ST
       22,60 €
  • WHAT10362010
    Universal indicator papers, non bleeding strips
    Universal indicator papers, non bleeding strips
    2,0 - 9,0
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 22,60 €
    22,60 €
    1 * 100 ST
       22,60 €