Order Entry
Belgium
Orders LinkContactUsLinkComponent
Furosemide, Reference Standards

Furosemide, Reference Standards

Supplier:   USP
Furosemide, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1287008.
  • 1287020.
  • 1287030.
  • Description:
  • Furosemide
  • Furosemide Related Compound A
  • Furosemide Related Compound B
  • Synonym(s):
  • 2-Chloro-4-N-furfurylamino-5-sulfamoylbenzoic Acid
  • 4-Chloro-5-sulfamoylanthranilic Acid
  • CAS No.:
  • 54-31-9
  • 4818-59-1
  • 3086-91-7
  • Formula:
  • C₁₂H₁₀ClN₂O₅S
  • C₁₂H₁₁ClN₂O₅S
  • C₇H₇ClN₂O₄S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1287008.
    Furosemide
    Furosemide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 391,00 €
    391,00 €
    1 * 200 mg
       391,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1287008.
    Description
    Furosemide
    Synonym(s)
    CAS No.
    54-31-9
    Formula
    C₁₂H₁₀ClN₂O₅S
  • 1287020.
    Furosemide Related Compound A
    Furosemide Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 240,00 €
    1 240,00 €
    1 * 50 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1287020.
    Description
    Furosemide Related Compound A
    Synonym(s)
    2-Chloro-4-N-furfurylamino-5-sulfamoylbenzoic Acid
    CAS No.
    4818-59-1
    Formula
    C₁₂H₁₁ClN₂O₅S
  • 1287030.
    Furosemide Related Compound B
    Furosemide Related Compound B
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 1 150,00 €
    1 150,00 €
    1 * 100 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1287030.
    Description
    Furosemide Related Compound B
    Synonym(s)
    4-Chloro-5-sulfamoylanthranilic Acid
    CAS No.
    3086-91-7
    Formula
    C₇H₇ClN₂O₄S
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.