Order Entry
Belgium
ContactUsLinkComponent
M. tuberculosis Recombinant NAD Synthetase (from E. coli)
M. tuberculosis Recombinant NAD Synthetase (from E. coli)
Catalog # PRSI90-366
Supplier:  ProSci Inc.
CAS Number:  
M. tuberculosis Recombinant NAD Synthetase (from E. coli)
Catalog # PRSI90-366
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    M. tuberculosis
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    NAD Synthetase
  • Enzyme activity:
    ~0.1 U/mg protein. (Ammonium or glutamine as nitrogen source). One unit is defined as the amount of enzyme that synthesizes 1umol NAD per min.
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Liquid, In 20 mM TRIS-HCl, pH 7.8.

Specifications

About this item

NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalysing the final step of both pathways. Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It can use both glutamine or ammonia as a nitrogen source. Catalytic activity: ATP + deamido-NAD+ + L-glutamine + H₂O = AMP + diphosphate + NAD+ + L-glutamate.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.