Order Entry
Belgium
ContactUsLinkComponent
 
M. tuberculosis Recombinant NAD Kinase (from E. coli)
  PRSI90-365
 :  
M. tuberculosis Recombinant NAD Kinase (from E. coli)
  PRSI90-365
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    M. tuberculosis
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    NAD Kinase
  • Enzyme activity:
    ~1.2 U/mg protein. (Substrate: NAD (+ ATP))
    ~3.8 U/mg protein. (Substrate: NAD (+ poly(P))
    One unit is defined as the amount of enzyme that synthesizes 1umol nicotinamide adenine dinucleotide phosphate (NADP) per min.
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Liquid, In 10 mM TRIS at pH 8.0, containing 1 mM MgCl₂, 0.5 mM EDTA, 1 mM DTT and 1 mM PMSF.

 

 

NAD kinase catalyses the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis. NAD kinase from Mycobacterium tuberculosis is not ATP-dependent but has a broader substrate specificity than human NAD kinase. It can economically produce NADP+ by using other nucleoside triphosphates as well as inorganic polyphosphate as a source of phosphorus. Was identified as a high-confidence drug target. Catalytic activity: ATP + NAD+ = ADP + NADP+.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.