Order Entry
Belgium
ContactUsLinkComponent
Human recombinant NAPRTase (from E. coli)
Human recombinant NAPRTase (from E. coli)
Catalog # PRSI90-363
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant NAPRTase (from E. coli)
Catalog # PRSI90-363
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    NAPRTase
  • Enzyme activity:
    ~0,27 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1 umol nicotinic acid mononucleotide (NaMN) per min.
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Liquid, In 50 mM HEPES/KOH pH 7,5 containing 300 mM potassium chloride and 10 mM DTT.

Specifications

About this item

Human nicotinate phosphoribosyltransferase (NAPRTase) is localised in the cytoplasm and is involved in biological processes such as NAD biosynthetic and metabolic processes, nicotinamide metabolic process, nicotinate nucleotide salvage, response to oxidative stress and water-soluble vitamin metabolic process. It functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN) and is essential for NA to increase cellular NAD levels and prevent oxidative stress of the cells. It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.