Order Entry
Belgium
ContactUsLinkComponent
Human recombinant phosphoglucomutase-2 (from cells)
Human recombinant phosphoglucomutase-2 (from cells)
  PRSI92-619
 :  
Human recombinant phosphoglucomutase-2 (from cells)
  PRSI92-619
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Phosphoglucomutase-2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    70.5 kDa
  • Sequence:
    Met1-Asp612
  • Formulation:
    Lyophilized from a 0,2 µm filtered solution of 20 mM Tris,200 mM NaCl,pH 8,0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

 

 

Phosphoglucomutase-2 (PGM2) is a member of PGM family, which catalyses the inter-conversion of sugar phosphates and participates in anabolic and catabolic reactions. When cells are grown in glucose, PGM catalyses the conversion of glucose-6-phosphate to glucose-1-phosphate an important precursor required for the synthesis of UDP glucose and trehalose. PGM2 catalyses the conversion of the nucleoside breakdown products ribose-1-phosphate and deoxyribose-1-phosphate to the corresponding 5-phosphopentoses, and it may also catalyse the interconversion of glucose-1-phosphate and glucose-6-phosphate. But this protein has low glucose 1,6-bisphosphate synthase activity.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.