Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Selenophosphate synthase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:43.9 kD
- Sequence:Met1-Ser392
- Formulation:Supplied as a 0.2 µm filtered solution of 25 mM Tris.HCl, pH 7.3, 100 mM glycine, 10% Glycerol. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
Selenophosphate synthetase 1 (SEPHS1) belongs to the selenophosphate synthase 1 family, Class II subfamily. It has four different isoforms by alternative splicing. Isoform 1 and isoform 2 are gradually expressed during the cell cycle until G2/M phase and then decreased, which Isoform 3 is gradually expressed during the cell cycle until S phase and then decreased. SEPHS1 can be activated by phosphate ions and by potassium ions. It can synthesise synthesises selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesise selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.