Human recombinant D-tyrosyl-tRNA(Tyr) deacylase 1 (from cells)
Catalog # PRSI92-377
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:D-tyrosyl-tRNA(Tyr) deacylase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:24.5 kD
- Sequence:Met1-Pro209
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
D-tyrosyl-tRNA(Tyr) deacylase 1(DTD1) belongs to the DTD family, and expressed in many adult and fetal tissues such as testis, ovary, spleen in adult and fetal brain. It is a nucleus and cytoplasm located protein, and is preferentially phosphorylated in cells arrested early in S phase. DTD1 is an ATPase involved in DNA replication, it may facilitate loading of CDC45 onto pre-replication complexes. The protein may hydrolyse D-tyrosyl-tRNA(Tyr) into D-tyrosine and free tRNA(Tyr), a possible defense mechanism against a harmful effect of D-tyrosine.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.