Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Polypeptide GalNac transferase 3 (from cells)
Human recombinant Polypeptide GalNac transferase 3 (from cells)
Catalog # PRSI92-355
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Polypeptide GalNac transferase 3 (from cells)
Catalog # PRSI92-355
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Polypeptide GalNac Transferase 3
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    69.1 kD
  • Sequence:
    Gln38-Asp633
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Polypeptide N-acetylgalactosaminyltransferase 3(GALNT3) belongs to the glycosyltransferase 2 family and galNAc-T subfamily. It expressed in organs that contain secretory epithelial glands and it highly expressed in pancreas, skin, kidney and testis. There are two conserved domains in the glycosyltransferase region: the N-terminal domain (domain A, also called GT1 motif), which is probably involved in manganese coordination and substrate binding and the C-terminal domain (domain B, also called Gal/GalNAc-T motif), which is probably involved in catalytic reaction and UDP-Gal binding .This protein plays a major role in regulating phosphate levels within the body (phosphate homeostasis). Among its many functions, phosphate plays a critical role in the formation and growth of bones in childhood and helps maintain bone strength in adults.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.