Human recombinant malate dehydrogenase (MDH) (from cells)
Catalog # PRSI92-353
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Malate dehydrogenase (MDH)
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:34 kD
- Sequence:Ala25-Lys338
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the lyophilized protein in dd H₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
Malate dehydrogenase, mitochondrial is a 338 amino acids protein that belongs to the LDH/MDH superfamily. MDH type 1 family. MDH2 catalyses the reversible oxidation of malate to oxaloacetate, utilising the NAD/NADH cofactor system in the citric acid cycle. MDH2 is localised to the mitochondria and takes part in the malate-aspartate shuttle that functions in the metabolic coordination between cytosol and mitochondria. MDH2 is highly expressed in the adrenal system, small intestine, heart and pancreas.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.