Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Aminoacylase-3 (from E. coli)
Human recombinant Aminoacylase-3 (from E. coli)
Catalog # PRSI92-310
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Aminoacylase-3 (from E. coli)
Catalog # PRSI92-310
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Aminoacylase-3
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37.4 kD
  • Sequence:
    Met1-Ser319
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris,100 mM NaCl,1 mM DTT, 10% Glycerol,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Aspartoacylase 3, also known as ACY3, N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming), Acylase III, Aminoacylase-3, Aspartoacylase-2, Aspartoacylase-2, HCV core-binding protein 1 and ASPA2, is a member of the Aspartoacylase subfamily. ACY3 plays an important role in deacetylating mercapturic acids in kidney proximal tubules and acts on N-acetyl-aromatic amino acids.ACY3 is located in the cytoplasm of S2 and S3 proximal tubules and the apical domain of S1 proximal tubules. ACY3 protein is also expressed at low levels in stomach, testis, heart, brain, lung and liver, and may function as an HCV (Hepatitis C virus) core binding protein.

Fusion-Tag: N-6 His tag.

This recombinant protein can be used for biological assays. For research use only.