Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant Dipeptidyl-peptidase 3 (from E. coli)
Human Recombinant Dipeptidyl-peptidase 3 (from E. coli)
Catalog # PRSI92-263
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant Dipeptidyl-peptidase 3 (from E. coli)
Catalog # PRSI92-263
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Dipeptidyl-peptidase 3
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    84.8 kD
  • Sequence:
    Met1-Ala737
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Dipeptidyl peptidase 3(DPP3), is a member of the S9B family in clan SC of the serine proteases. DPP3 has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH). It releases an N-terminal dipeptide from a peptide comprising four or more residues, with broad specificity and also acts on dipeptidyl 2-naphthylamides.Increased activity of this protein has a relationship with endometrial and ovarian cancers.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.