Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Carbonic Anhydrase 14 (from E. coli)
Human recombinant Carbonic Anhydrase 14 (from E. coli)
Catalog # PRSI92-149
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Carbonic Anhydrase 14 (from E. coli)
Catalog # PRSI92-149
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Carbonic Anhydrase 14
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    32.8 kD
  • Sequence:
    Gly19-Met290
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Carbonic Anhydrase 14 (CA14) belongs to the Alpha-Carbonic Anhydrase family. It is highly expressed in all parts of the central nervous system and lowly expressed in adult liver, heart, small intestine, colon, kidney, urinary bladder, and skeletal muscle. CA14 along with other Carbonic Anhydrases (CAs) participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. CA14 is predicted to be a type I membrane protein and catalyses the reversible hydration of carbon dioxide.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.