Human recombinant ASM-like phosphodiesterase 3a (from Cells)
Catalog # PRSI91-932
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:ASM-like phosphodiesterase 3a
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:49.9 kD
- Sequence:Leu23-Tyr453
- Formulation:Lyophilized from a 0.2 µm filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Acid sphingomyelinase-like phosphodiesterase 3a (SMPDL3A ) is a novel liver X receptor (LXR) -regulated gene, with an LXR response element within its promoter. The induction of SMPDL3A is LXR-dependent and is restricted to human blood cells with no induction observed in mouse cellular systems. LXR function as physiological sensors of cholesterol metabolites (oxysterols), regulating key genes involved in cholesterol and lipid metabolism. LXRs have been extensively studied in both human and rodent cell systems, revealing their potential therapeutic value in the contexts of atherosclerosis and inflammatory diseases.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.