Human recombinant beta-1,4-Galactosyltransferase 3 (from Cells)
PRSI91-863
:
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:beta-1,4-Galactosyltransferase 3
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:41.5 kD
- Sequence:Arg32-His393
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,2 mM MgCl₂, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Beta-1,4-galactosyltransferase 3 (B4GALT3) belongs to the glycosyltransferase 7 family. It is responsible for the synthesis of complex-type N-linked oligosaccharides in many glycoproteins as well as the carbohydrate moieties of glycolipids. It is highest expression in placenta, prostate, testis, ovary, intestine and muscle, and in fetal brain.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.