Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant prostaglandin-D2 Synthase (from cells)
  PRSI91-757
 :  
Human recombinant prostaglandin-D2 Synthase (from cells)
  PRSI91-757
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Prostaglandin-D2 Synthase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19.7 kD
  • Sequence:
    Ala23-Gln190
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Prostaglandin-H2 D-Isomerase (PTGDS) belongs to the Lipocalin family of calycin superfamily. PTGDS is preferentially expressed in the brain. PTGDS catalyses the conversion of PGH2 to PGD2, a prostaglandin involved in smooth muscle contraction/relaxation and a potent inhibitor of platelet aggregation. PTGDS is involved in a variety of CNS functions, such as sedation, REM sleep and PGE2-induced allodynia, and may have an anti-apoptotic role in oligodendrocytes. PTGDS binds small non-substrate lipophilic molecules and may act as a scavenger for harmful hydrophopic molecules and a secretory retinoid and thyroid hormone transporter. It possibly participates in development and maintenance of the blood-brain, blood-retina, blood-aqueous humor, blood-testis barrier, the central nervous system and male reproductive system.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.