Human recombinant CaM Kinase I (from Cells)
Catalog # PRSI91-691
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:CaM Kinase I
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:42.3 kD
- Sequence:Met1-Leu370
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Calcium/Calmodulin-Dependent Protein Kinase Type 1 (CAMK1) belongs to the protein kinase superfamily, CAMK Ser/Thr protein kinase family, and CaMK subfamily. CAMK1 contains one protein kinase domain and widely expressed. CAMK1 is phosphorylated by CaMKK1 and CaMKK2 on Thr-177. CAMK1 regulates transcription activators activity, cell cycle, hormone production, cell differentiation, actin filament organisation, and neurite outgrowth. CAMK1 plays a role in K+ and ANG2-mediated regulation of the aldosterone synthase (CYP11B2) to produce aldosterone in the adrenal cortex.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.