Order Entry
Belgium
ContactUsLinkComponent
Human recombinant ER alpha-1,2-Mannosidase (from cells)
Human recombinant ER alpha-1,2-Mannosidase (from cells)
Catalog # PRSI91-681
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ER alpha-1,2-Mannosidase (from cells)
Catalog # PRSI91-681
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    ER alpha-1,2-Mannosidase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    68.7 kD
  • Sequence:
    Asp106-Ala699
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 50 mM TrisHCL,10 mM reduced Glutathione,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Endoplasmic Reticulum Mannosyl-Oligosaccharide 1,2- alpha-Mannosidase (MAN1B1) belongs to the glycosyl hydrolase 47 family. MAB1B1 is a single-pass type II membrane protein and widely expressed in many tissues. MAB1B1 is involved in glycoprotein quality control targeting of misfolded glycoproteins for degradation. MAB1B1 can be inhibited by both 1-deoxymannojirimycin (dMNJ) and kifunensine. Defects in MAN1B1 are the cause of mental retardation autosomal recessive type 15 (MRT15). Mental retardation is characterised by significantly below average general intellectual functioning, it is also associated with impairments in adaptative behavior and manifested during the developmental period.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.