Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Carboxypeptidase A2 (from Cells)
Human recombinant Carboxypeptidase A2 (from Cells)
Catalog # PRSI91-529
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Carboxypeptidase A2 (from Cells)
Catalog # PRSI91-529
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Carboxypeptidase A2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    45.91 kD
  • Sequence:
    Leu17-Tyr417
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Carboxypeptidase A2 (CPA) is a secreted pancreatic procarboxy-peptidase that cleaves the C-terminal amide or ester bond of peptides that have a free C-terminal carboxyl group. The hydrolytic action of CPA2 was identified with a preference towards long substrates with aromatic amino acids in their C-terminal end, particularly tryptophan. CPA2 comprises a signal peptide, a pro region and a mature chain, and can be activated after cleavage of the pro peptide. Three different forms of human pancreatic procarboxypeptidase A have been isolated, and the A1 and A2 forms are always secreted as monomeric proteins with different biochemical properties. In contrast to procarboxypeptidase B which was always secreted by the pancreas as a monomer, procarboxypeptidase A occurs as a monomer and/or associated to one or two functionally different proteins, such as zymogen E, and is involved in zymogen inhibition.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.