- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:DMQ hydroxylase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:21.34 kD
- Sequence:Ser37-Leu217
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Ubiquinone Biosynthesis Protein COQ7 Homolog (COQ7) is a mitochondrion inner membrane and peripheral membrane protein which belongs to the COQ7 family. It is expressed dominantly in heart and skeletal muscle. COQ7 is synthesised as a preprotein that is imported into the mitochondrial matrix, where the sequence is cleaved off and the mature protein becomes loosely associated with the inner membrane. COQ7 is involved in lifespan determination in ubiquinone-independent manner and also involved in ubiquinone biosynthesis. COQ7 is potential central metabolic regulator. Human COQ7 protein contains 179 amino acids, is mostly helical, and contains an alpha-helical membrane insertion. It has been shown that mutations in the gene are associated with increased life span. Defects of the gene slow down a variety of developmental and physiological processes, including the cell cycle, embryogenesis, post-embryonic growth, rhythmic behaviors and aging.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.