Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Chitinase 3-like 2 (from cells)
Human recombinant Chitinase 3-like 2 (from cells)
Catalog # PRSI91-523
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Chitinase 3-like 2 (from cells)
Catalog # PRSI91-523
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Chitinase 3-Like 2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    41.94 kD
  • Sequence:
    Tyr27-Leu390
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Chitinase 3-Like Protein 2 (CHI3L2) is a 39 kDa secreted glycoprotein which belongs to the glycosyl hydrolase 18 family and the most closely related to human cartilage glycoprotein 39, which promotes the growth of human synovial cells as well as skin and fetal lung fibroblasts. Highest expression of CHI3L2 is in chondrocytes, followed by synoviocytes, lung and heart. It is not detected in spleen, pancreas, and liver. CHI3L2 may also be expressed in developing brain and placenta. Increased levels of CHI3L2 have been demonstrated in synovial fluids of patients with rheumatoid or osteoarthritis as well as in some other pathologies and in malignant tumors, particularly in glioblastomas. CHI3L2 may bind glycan structure with high affinity, but not heparin. It has has no chitotriosidase activity, but is likely to bind some type of glycan.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.