Human recombinant Carnosine Dipeptidase 1 (from Cells)
Catalog # PRSI91-392
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Carnosine Dipeptidase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:54.9 kD
- Sequence:Pro28-His507
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Carnosine Dipeptidase 1 (CNDP1) belongs to the M20 metalloprotease family. CNDP1 is specifically expressed in the brain, serum and adult nervous central system. It is identified as human carnosinase. CNDP1 contains trinucleotide (CTG) repeat length polymorphism in the coding region and is inhibited by the metal chelator 1,10-o-phenantrolin. In addition, CNDP1 can hydrolyse the beta-Ala|-His dipeptide (carnosine), anserine, Xaa-|-His dipeptides and other dipeptides including homocarnosine. CNDP1 deficiency has been associated with homocarnosinosis disease.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.