Human recombinant Carbonic Anhydrase 10 (from Cells)
Catalog # PRSI91-285
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Carbonic Anhydrase 10
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:32.82 kD
- Sequence:Gln22-Asn300
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Carbonic Anhydrase X (CA10) belongs to CA family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes such as respiration, renal tubular acidification and bone resorption. While CA10 is a secreted protein without Carbonic Anhydrase activity (i.e., the reversible hydration of CO2) due to point mutations in the zinc binding site, it has esterase activity. The human and mouse CA10 are expressed in the brain, indicating that they may play a role in brain development.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.