Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant Aldo-Keto Reductase 1C3 (from E. coli)
N° de catalogue PRSI91-275
Fournisseur:  ProSci Inc.
Numéro CAS:  
Human recombinant Aldo-Keto Reductase 1C3 (from E. coli)
N° de catalogue PRSI91-275
Fournisseur:  ProSci Inc.
Numéro CAS:  

Spécifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Aldo-Keto Reductase 1C3
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    36.74 kD
  • Sequence:
    Met1-Tyr323
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 100 mM NaCl, 1 mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Spécifications

À propos de cet article

Aldo-Keto Reductase Family 1 Member C2 (AKR1C2) plays a role in concert with the 5- alpha/5- beta -Steroid Reductases to convert Steroid hormones into the 3- alpha/5- alpha and 3- alpha/5- beta -Tetrahydrosteroids. AKR1C2 catalyses the inactivation of the most potent androgen 5- alpha-Dihydrotestosterone (5- alpha-DHT) to 5- alpha-Androstane-3- alpha, 17- beta -diol (3- alpha-diol).

This recombinant protein can be used for biological assays. For research use only.