Human recombinant PPIase H (from E. coli)
Catalog # PRSI91-233
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:PPIase H
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:21.4 kD
- Sequence:Met1-Met177
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Peptidyl-Prolyl Cis-Trans Isomerase H (PPIH) belongs to the Cyclophilin-type PPIase family that accelerate the folding of proteins. PPIases can catalyze the cis-trans isomerisaion of Proline Imidic peptide bonds in oligopeptides. PPIH participates in pre-mRNA splicing. It is a specific component of the complex that includes pre-mRNA processing factors PRPF3, PRPF4, and PRPF18, as well as U4/U5/U6 tri-snRNP. In addition, PPIH has PPIase activity and may play a role as a chaperone mediating the interactions between different proteins inside the spliceosome.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.