Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant B7-H4 (from CHO cells)
Mouse recombinant B7-H4 (from CHO cells)
Catalog # PRSI90-371
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant B7-H4 (from CHO cells)
Catalog # PRSI90-371
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological activity:
    Measured by its ablility to inhibit anti-CD3-induced proliferation of stimulated mouse T cells.
  • Protein synonyms:
    Immune Costimulatory Protein B7-H4|V-set Domain-containing T Cell Activation Inhibitor 1|B7h.5|Protein B7S1|VTCN1|T Cell Costimulatory Molecule B7x
  • Protein/peptide name:
    B7-H4
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    29-258
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

B7-H4 is a B7 family member that negatively regulates T cell immunity by inhibiting T cell proliferation, cytokine production and cell cycle progression. In vitro, B7-H4 inhibits CD4+ and CD8+ T cell proliferation, cytokine production and generation of alloreactive cytotoxic T cells (CTLs). In vivo, blockade of endogenous B7-H4 by specific monoclonal antibody promotes T cell responses. B7-H4 ia an important negative regulator of innate immunity through growth inhibition of neutrophils. B7-H4 is expressed on some tumour cancer cells. The role of B7-H4 in tumour progression may be to transform precancerous cells and then protect them from immunosurveillance.

  • This recombinant proteins is for research use only.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.