Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant TNF-like 1 (from E. coli)
Mouse recombinant TNF-like 1 (from E. coli)
Catalog # PRSI92-684
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant TNF-like 1 (from E. coli)
Catalog # PRSI92-684
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Tumor Necrosis Factor Ligand Superfamily Member 15|TL1A|Vascular Endothelial Cell Growth Inhibitor|VEGI|TNFSF15|TL1|TNF Ligand-Related Molecule 1
  • Protein/peptide name:
    TNF-like 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    20 kD
  • Sequence:
    Ile76-Leu252
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Tumour Necrosis Factor Ligand Superfamily Member 15 (TNFSF15) is a new member of the tumour necrosis factor family. TNFSF15 is predominantly an endothelial cell-specific gene, and recombinant TNFSF15 is a potent inhibitor of endothelial cell proliferation, angiogenesis and tumour growth. TNFSF15 exerts two activities on endothelial cells: early G1 arrest of G0/G1-cells responding to growth stimuli and programmed cell death of proliferating cells. These activities are highly specific to endothelial cells. TNFSF15 is also able to regulate the expression of several important genes involved in angiogenesis. These findings are consistent with the view that TNFSF15 functions as an autocrine cytokine to inhibit angiogenesis and stabilise the vasculature.

This recombinant protein can be used for biological assays. For research use only.