Order Entry
Belgium
ContactUsLinkComponent
Human recombinant ULBP-1 (from cells)
Human recombinant ULBP-1 (from cells)
Catalog # PRSI92-572
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ULBP-1 (from cells)
Catalog # PRSI92-572
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    N2DL-1|UL16-binding protein 1|NKG2DL|ALCAN-beta|NKG2DL1|Retinoic acid early transcript 1I|NKG2D ligand 1|ULBP1
  • Protein/peptide name:
    ULBP-1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.3 kD
  • Sequence:
    Gly26-Pro215
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

ULBP1, also known as RAET1I and NKG2DL1, is a member of the ULBP/RAET1 gene family. ULBP1 plays an important role in immune responses, especially in cancer and infectious diseases, and is well-known to bind to NKG2D together with at least ULBP 2 and 3. These proteins are distantly related to major histocompatibility class I (MHC I) molecules, possessing the alpha 1 and alpha 2 Ig-like domains, but lacking the alpha 3 domain. Unlike MHC Class I, they have no capacity to bind peptide or interact with beta2-microglobulin. It can activate multiple signallling pathways in primary NK cells, gamma delta T cells, and CD8+ alpha beta T cells, resulting in the production of cytokines and chemokines.ULBP1 is expressed in wide range of tissues including heart, brain, lung, liver, bone marrow and some tumour cells, T-cells, B-cells, As an unconventional member of the MHC class I family, ULBP1 is able to interact with soluble CMV glycoprotein UL16 in CMV infected cells. The interaction with UL16 blocked the interaction with the NKG2D receptor, and thus might escape the immune surveillance. Furthermore, UL16 also causes ULBP1 to be retained in the ER and cis-Golgi apparatus so that it does not reach the cell surface. The ULBP1 regulation may have implications for development of new therapeutic strategies against cancer cells.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.