Order Entry
Belgium
ContactUsLinkComponent
Human recombinant UBE2B (from E. coli)
Human recombinant UBE2B (from E. coli)
Catalog # PRSI92-111
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant UBE2B (from E. coli)
Catalog # PRSI92-111
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    HR6B|Ubiquitin-Conjugating Enzyme E2-17 kDa|UBE2B|RAD6B|Ubiquitin Carrier Protein B|Ubiquitin-Protein Ligase B|hHR6B|Ubiquitin-Conjugating Enzyme E2 B|RAD6 Homolog B
  • Protein/peptide name:
    UBE2B
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Met1-Ser152
  • Formulation:
    Supplied as a 0.2 um filtered solution of 10mM HEPES, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Ubiquitin-Conjugating Enzyme E2 B (UBE2B) belongs to the ubiquitin-conjugating enzyme family. UBE2B can catalyze the covalent attachment of ubiquitin to other proteins, and is essential for the multi-ubiquitination and degradation of N-end rule substrates. UBE2B is indispensability for postreplication repair of UV-damaged DNA and may be involved in neurite outgrowth.Additionally, UBE2B may have a role in sepsis-induced muscle protein proteolysis and cancer-induced cachexia.

  • This recombinant protein can be used for biological assays. For research use only.