Human recombinant Annexin A10 (from E. coli)
Catalog # PRSI92-062
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Annexin-10|ANXA10|ANX14|Annexin-14|Annexin A10
- Protein/peptide name:Annexin A10
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:37.5 kD
- Sequence:Met1-Tyr324
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
Annexin A10 (ANXA10) contains four Annexin repeats and is a member of the Annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. It is reported that ANXA10 may be a clinical relevant marker for predicting outcome in both early and advanced stages of bladder cancer.
- This recombinant protein can be used for biological assays. For research use only.