Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Estrogen Receptor alpha (from E. coli)
Human recombinant Estrogen Receptor alpha (from E. coli)
Catalog # PRSI91-978
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Estrogen Receptor alpha (from E. coli)
Catalog # PRSI91-978
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    ER|ER alpha|Nuclear Receptor Subfamily 3 Group A Member 1|NR3A1|Estrogen Receptor|ESR|ESR1|Estradiol Receptor|ER-Alpha
  • Protein/peptide name:
    Estrogen Receptor alpha
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    14.38 kD
  • Sequence:
    Met1-Gln116
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Estrogen Receptor is a major ligand-activated transcription factor belonging to the nuclear hormone receptor superfamily. Estrogen Receptor is composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but they also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5' UTRs and use different promoters.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag