Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant CD14 (from Cells)
Human Recombinant CD14 (from Cells)
Catalog # PRSI91-288
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant CD14 (from Cells)
Catalog # PRSI91-288
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Myeloid Cell-Specific Leucine-Rich Glycoprotein|Monocyte Differentiation Antigen CD14|CD14
  • Protein/peptide name:
    CD14
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    36.81 kD
  • Sequence:
    Thr20-Cys352
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

CD14 is a cell surface glycoprotein that is preferentially expressed on monocytes/macrophages. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a pattern recognition receptor that binds lipopolysaccharides (LPS) and a variety of ligands derived from different microbial sources. The binding of CD14 with LPS is catalyzed by LPS binding protein (LBP). Toll like receptors have also been implicated in the transduction of CD14-LPS signals. Soluble CD14 can be released from the cell surface by phosphatidyinositolspecific phospholipase C and has been detected in serum and body fluids. High concentrations of soluble CD14 have been shown to inhibit LPS mediated responses. However, soluble CD14 can also potentiate LPS response in cells that do not express cell surface CD14.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag