Order Entry
Belgium
ContactUsLinkComponent
Human recombinant PKI-beta (from E. coli)
Human recombinant PKI-beta (from E. coli)
  PRSI91-227
 :  
Human recombinant PKI-beta (from E. coli)
  PRSI91-227
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    PKIB|PRKACN2|PKI-beta|cAMP-Dependent Protein Kinase Inhibitor Beta
  • Protein/peptide name:
    PKI-beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10.6 kD
  • Sequence:
    Met1-Lys78
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 1mM DTT, 20% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

cAMP-Dependent Protein Kinase Inhibitor beta (PKI- beta ) is a member of the PKI family. It has been shown that PKI- beta is an extremely potent competitive inhibitor of cAMP-dependent protein kinase activity; this protein interacts with the catalytic subunit of the enzyme after the cAMP-induced dissociation of its regulatory chains.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag