Order Entry
Belgium
ContactUsLinkComponent
Human recombinant NHP2L1 (from E. coli)
Human recombinant NHP2L1 (from E. coli)
Catalog # PRSI91-220
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant NHP2L1 (from E. coli)
Catalog # PRSI91-220
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    U4/U6.U5 tri-snRNP 15.5 kDa Protein|NHP2L1|OTK27|High Mobility Group-like Nuclear Protein 2 Homolog 1|hSNU13|NHP2-like Protein 1|SNU13 Homolog
  • Protein/peptide name:
    NHP2L1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.3 kD
  • Sequence:
    Met1-Val128
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 600mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

NHP2-Like Protein 1 (NHP2L1) is a member of the ribosomal protein L7Ae family. NHP2L1 proteinis limited to the nucleus, primarily focused in the dense fibrillar component of the nucleolus. NHP2L1 has been shown to interact with RAD17and PRPF31. The protein undergoes a conformational change upon RNA-binding. NHP2L1 binds to the 5-stem-loop of U4 snRNA and may play a role in the late stage of spliceosome assembly, prior to step I of splicing catalysis.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag