Order Entry
Belgium
ContactUsLinkComponent
Human recombinant E Selectin1 (from HEK293 cells)
Human recombinant E Selectin1 (from HEK293 cells)
Catalog # PRSI96-168
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant E Selectin1 (from HEK293 cells)
Catalog # PRSI96-168
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    200 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    E-Selectin|RP1-117P20.2|ELAM1|ESEL|LECAM2|CD62E|SELE
  • Protein/peptide name:
    E Selectin1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    60 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

E-Selectin is also known as CD62 antigen-like family member E (CD62E), endothelial-leukocyte adhesion molecule 1 (ELAM-1), or leukocyte-endothelial cell adhesion molecule 2 (LECAM2), a member of the Selectin family, is a 107-115 kDa cell surface glycoprotein. It is transiently expressed on vascular endothelial cells in response to IL-1β and TNFα. E selectin has a cassette structure: an N-terminal, C-type lectin domain, an EGF (epidermal-growth-factor)-like domain, 6 Sushi domain (SCR repeat) units, a transmembrane domain (TM) and an intracellular cytoplasmic tail (cyto). During inflammation, E-selectin plays an important part in recruiting leukocytes to the site of injury. The local release of cytokines IL-1 and TNF-α by damaged cells induces the over-expression of E-selectin on endothelial cells of nearby blood vessels. E-selectin mediates the adhesion of tumor cells to endothelial cells, by binding to E-selectin ligands expressed by neutrophils, monocytes, eosinophils, memory-effector T-like lymphocytes, natural killer cells or cancer cells. Furthermore, a number of studies have reported that levels of E-Selectin may be elevated in subjects with a variety of pathological conditions.