Human recombinant RANTES (from E. coli)
Catalog # PRSI40-567
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:Biological activity was determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10.0 ng/mL. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
- Protein synonyms:EoCP|RANTES|C-C motif chemokine 5|SCYA5|SIS-delta|eoCP|TCP228|D17S136E|SISd
- Protein/peptide name:RANTES
- Purity:>98%
- Formulation:Lyophilized
Specifications
About this item
RANTES is a CC-chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant human RANTES is a 7.8 kDa protein containing 68 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
- Store in working aliquots at -20°C. Avoid freeze/thaw cycles.