Order Entry
Belgium
ContactUsLinkComponent
Human recombinant PPIL1 (from E. coli)
Human recombinant PPIL1 (from E. coli)
Catalog # PRSIXW-RP3204
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant PPIL1 (from E. coli)
Catalog # PRSIXW-RP3204
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    UNQ2425/PRO4984|hCyPX|Peptidyl-prolyl cis-trans isomerase-like 1|CGI-124|CYPL1|PPIase|Rotamase PPIL1
  • Protein/peptide name:
    PPIL1
  • Purity:
    >95%
  • Molecular weight:
    19.1 kDa (Calculated)
  • Tested applications:
    ELISA, Western Blot, MS

Specifications

About this item

This protein is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.

  • This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.