Order Entry
Belgium
ContactUsLinkComponent
Human DELE peptide
Human DELE peptide
Catalog # PRSI6919P
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human DELE peptide
Catalog # PRSI6919P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 amino acid peptide near the amino terminus of human DELE
  • Protein/peptide name:
    DELE Peptide
  • Tested applications:
    Blocking

Specifications

About this item

DELE peptide is used for blocking the activity of DELE antibody.