Order Entry
Belgium
ContactUsLinkComponent
 
Human RPA interacting protein peptide (17 aa near C-terminus)
  PRSI4013P
 :  
Human RPA interacting protein peptide (17 aa near C-terminus)
  PRSI4013P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 aa near C-terminus
  • Protein/peptide name:
    RPA interacting protein Peptide (17 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

RPA interacting protein peptide is used for blocking the activity of RPA interacting protein antibody.