Order Entry
Belgium
ContactUsLinkComponent
SARS E protein SARS E protein peptide (10 aa near N-terminus)
SARS E protein SARS E protein peptide (10 aa near N-terminus)
Catalog # PRSI3531P
Supplier:  ProSci Inc.
CAS Number:  
SARS E protein SARS E protein peptide (10 aa near N-terminus)
Catalog # PRSI3531P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    SARS E protein
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    10 aa near N-terminus
  • Protein/peptide name:
    SARS E protein Peptide (10 aa near N-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

SARS envelope peptide is used for blocking the activity of the SARS envelope antibody.