Order Entry
Belgium
ContactUsLinkComponent
SARS spike protein SARS Spike protein peptide (14 aa near N-terminus)
SARS spike protein SARS Spike protein peptide (14 aa near N-terminus)
Catalog # PRSI3219P
Supplier:  ProSci Inc.
CAS Number:  
SARS spike protein SARS Spike protein peptide (14 aa near N-terminus)
Catalog # PRSI3219P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    SARS Spike protein
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    14 aa near N-terminus
  • Protein/peptide name:
    SARS Spike protein Peptide (14 aa near N-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

SARS Spike peptide is used for blocking the activity of the SARS Spike antibody.