ContactUsLinkComponent
Portable/wall group lockout box, Prinzing
Portable/wall group lockout box, Prinzing
Catalog # BRDY046134
Supplier:  Brady
undefined
Portable/wall group lockout box, Prinzing
Catalog # BRDY046134
Supplier:  Brady
Specifications
  • Type:
    Portable/wall group lockout box
  • Colour:
    Yellow
  • W×D×H:
    354,5×73,4×151 mm
  • Pk:
    1
Specifications
About this item
This box accommodates 8 to 10 locks (depending on lock size).
  • Made of yellow powder-coated metal (mild sheet steel)
  • Can be used alone or in combination with Industrial Strength lockout station
Ordering information: Contents not included.