Anti-CCND1 Rabbit Polyclonal Antibody
N° de catalogue PRSI80-035
Fournisseur: ProSci Inc.
Spécifications
- Antibody type:Primary
- Antigen name:Cyclin D1
- Antigen symbol:CCND1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:595
- Antigen synonyms:BCL1|PRAD1 oncogene|PRAD1|BCL-1 oncogene|G1/S-specific cyclin-D1|B-cell lymphoma 1 protein|U21B31|B-cell CLL/lymphoma 1|D11S287E
- Modification:phospho-Ser90
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:31 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:-20°C
- Immunogen:Cyclin D1 (phospho Ser90) antibody was raised against a peptide sequence around phosphorylation site of serine 90 (F-L-S (p)-L-E) derived from Human Cyclin D1 .
- Purification:Purified by affinity-chromatography using epitope-specific peptide.
- Size:100 µg
- Pk:100 µG
Spécifications
À propos de cet article
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes 'Lys-48'-linked polyubiquitination. Mediates the selective degradation of short-lived and abnormal proteins. Functions in the E6/E6-AP-induced ubiquitination of p53/TP53. Mediates ubiquitination of PEX5 and auto-ubiquitination of STUB1, TRAF6 and TRIM63/MURF1. Ubiquitinates STUB1-associated HSP90AB1 in vitro. Lacks inherent specificity for any particular lysine residue of ubiquitin. Essential for viral activation of IRF3. Mediates polyubiquitination of CYP3A4.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: CCND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat