Anti-CD70 Rabbit Polyclonal Antibody
PRSI71-147
- Antigen name:CD70 molecule
- Antigen symbol:CD70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gorilla,Human
- Antigen synonyms:CD27LG|TNFSF7|CD27L
- Concentration:1 mg/mL
- Immunogen:Synthetic 17 amino acid peptide from internal region of human CD70.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
CD70 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 - 10 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CD70
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: