Anti-CARD16 Goat Polyclonal Antibody
PRSI45-417
- Antigen name:Caspase Recruitment Domain Family, Member 16
- Antigen symbol:CARD16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:COP1|COP|PSEUDO-ICE
- Concentration:500 ug/mL
- Immunogen:COP1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of COP1.
- Size:100 µg
- Pk:100 µG
COP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
COP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CARD16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human